Learn More
Hirschmann™ Individual Calibration Certificates for Volumetric Pipettes
Providing verification of an extremely high degree of volumetric accuracy for your pipetting needs.
Brand:  Hirschmann™ 134015027-9980104
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15655455
  • 167.00€ / Pack of 6
Estimated Shipment
22-09-2020
Add to basket
Description
Description
  • DAkkS German Calibration Service
  • For volumetric measuring devices
  • Internationally recognized
  • A valuable contribution to traceability and certification
Specifications
Specifications
Yes
DAkkS
For 4 mL pipettes
Individual Calibration Certificate