missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Indo-1 AM Calcium Sensor Dye
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
- Unbound Indo-1 has a peak emission at 475nm
- Upon binding calcium, peak emission shifts down to 410nm
- In flow cytometry this can be measured over time and can be represented as a ratio of the two emission wavelengths
- To optimize the ratio between the two emissions, unbound Indo-1 fluorescence should be collected using a filter above 485nm; bound Indo-1 fluorescence should be collected using a filter below 400nm
- Because the emission profile of Indo-1 is broad, multicolor flow analysis using eFluor ™ 450, eVolve™ 605 or eVolve™ 655 is not possible, however fluorochrome conjugated antibodies utilizing the 488nm or 633 nm are quite compatible with Indo-1
- Molecular weight: 1009.91g/mol
- Peak excitation: 346nm
- Peak emission: 475nm
- Reconstitute in high-quality, freshly opened DMSO
- Recommended concentration: 1 mM
- Once reconstituted, store at −20°C with dessicant and use within a short period of time. Avoid freeze-thawing
Specifications
Specifications
| Color | Blue |
| Detection Method | Fluorescence |
| For Use With (Application) | Immunohistochemistry (IHC), Flow Cytometry |
| For Use With (Equipment) | Fluorescence Microscope |
| Product Type | Dye |
| Emission | 346/475 |
| Product Line | eBioscience |
| Quantity | 1 mg |
For Research Use Only. Not for use in diagnostic procedures.
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu