missing translation for 'onlineSavingsMsg'
Learn More

Indole-6-carbonitrile, 98+%

Product Code. 11364536
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
11364536 250 mg 250mg
11344536 1 g 1g
11354536 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 11364536

Brand: Thermo Scientific Alfa Aesar L20099.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 15861-36-6 | C9H6N2 | 142.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15861-36-6
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.161
MDL Number MFCD00016732
InChI Key SZSZDBFJCQKTRG-UHFFFAOYSA-N
Synonym 6-cyanoindole, 6-indolecarbonitrile, indole-6-carbonitrile, 6-cyano-1h-indole, indole-6-cyano, 6-cyano-indole, 6-indole carbonitrile, pubchem7239, cyanoindole-6, 1h-indol-6-yl cyanide
PubChem CID 85146
IUPAC Name 1H-indole-6-carbonitrile
SMILES C1=CC(=CC2=C1C=CN2)C#N

Specifications

Melting Point 128°C to 132°C
Odor Odorless
Quantity 250 mg
Beilstein 116502
Sensitivity Light sensitive
Formula Weight 142.16
Percent Purity ≥98%
Chemical Name or Material Indole-6-carbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.