Specifications
| Certifications/Compliance | Yes |
| Product Type | Infrared-Thermometer |
| Max. Temperature (Metric) | 110°C |
| Min. Temperature (Metric) | -22°C |
| Temperature Range (Metric) | -22°C to +110 x 0.5°C |