Learn More
OHAUS™ Innoculating Turntable
Product Code. p-7158948 Shop All Ohaus Products
Click to view available options
Size:
114 x 76 mm (H x dia.)
32 x 150 mm (H x dia.)
Unit Size:
Each
Product Code. 15782459
Brand: OHAUS™ 30400046
Only null left
Add to basket
Add to basket
OHAUS™ Innoculating Turntable provides secure support for holding glassware at elevated positions.
  • OHAUS offers the largest selection of lab-frame kits, lab-lifts, rods and support stands in the industry to hold glassware safely
  • Standard lab-frames or customisable set-ups with options to fit any space or application
  • Select from 13 pre-configured lab-frame kits (connectors and feet included) to rods which can be purchased individually and combined with OHAUS accessories to build a customized frame
TRUSTED_SUSTAINABILITY
Specifications
Type Innoculating Turntable
Material Cast Iron
For Use With (Equipment) 100 mm Petri Dish
Description Clamp, Non-Electrical, CLN-INOTTS
Diameter (Metric) 114 mm
Height (Metric) 76 mm
Height (English) 3.00 in.
Size 114 x 76 mm (H x dia.)