Learn More
Abeba™ Insoles 3556
ESD dissipative insoles include antibacterial and antifungal properties due to active carbon.
Brand: Abeba™ 3556/40
Additional Details : Weight : 0.04000kg
Description
- Breathable textile
- Blue
- ESD dissipative according to DIN EN 61340
- Antistatic
- Areal pressure absorption, climatising, comfortable
- Benefit for joints, musculature, ligaments and spine
Specifications
Blue | |
Textile | |
40 | |
3556 |