Learn More
Abeba™ Insoles 3556
ESD dissipative insoles include antibacterial and antifungal properties due to active carbon.
Brand:  Abeba™ 3556/38
Additional Details : Weight : 0.04000kg
 View more versions of this product
Product Code. 15680138
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • Breathable textile
  • Blue
  • ESD dissipative according to DIN EN 61340
  • Antistatic
  • Areal pressure absorption, climatising, comfortable
  • Benefit for joints, musculature, ligaments and spine
Specifications
Specifications
Blue
Textile
38
3556