Learn More
Abeba™ Insoles 3573
ESD dissipative insoles provide shock absorption on all the plentar surfaces.
Brand: Abeba™ 3573/39
Additional Details : Weight : 0.02000kg
Description
- Breathable textile
- Gray
- ESD dissipative according to DIN EN 61340
- Antistatic
- Areal pressure absorption, climatising, comfortable
- Benefit for joints, musculature, ligaments and spine
Specifications
Gray | |
Textile | |
39 | |
3573 |