Learn More
Abeba™ Insoles 3573
ESD dissipative insoles provide shock absorption on all the plentar surfaces.
Brand:  Abeba™ 3573/45
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 15680268
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • Breathable textile
  • Gray
  • ESD dissipative according to DIN EN 61340
  • Antistatic
  • Areal pressure absorption, climatising, comfortable
  • Benefit for joints, musculature, ligaments and spine
Specifications
Specifications
Gray
Textile
45
3573