Learn More
Abeba™ Insoles 3574
ESD dissipative insoles provide shock absorption on all the plentar surfaces.
13.10€
Specifications
Color Gray
Material Textile
Style 3574
View More Specs
Products 14
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15620278
View Documents Promotion Details
Abeba™
3574/35
35 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15630278
View Documents Promotion Details
Abeba™
3574/36
36 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15640278
View Documents Promotion Details
Abeba™
3574/37
37 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15650278
View Documents Promotion Details
Abeba™
3574/38
38 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15660278
View Documents Promotion Details
Abeba™
3574/39
39 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15670278
View Documents Promotion Details
Abeba™
3574/40
40 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15680278
View Documents Promotion Details
Abeba™
3574/41
41 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15690278
View Documents Promotion Details
Abeba™
3574/42
42 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15600288
View Documents Promotion Details
Abeba™
3574/43
43 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15610288
View Documents Promotion Details
Abeba™
3574/44
44 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15620288
View Documents Promotion Details
Abeba™
3574/45
45 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15630288
View Documents Promotion Details
Abeba™
3574/46
46 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15640288
View Documents Promotion Details
Abeba™
3574/47
47 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15650288
View Documents Promotion Details
Abeba™
3574/48
48 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description
Description
  • Breathable textile
  • Gray
  • ESD dissipative according to DIN EN 61340
  • Antistatic
  • Areal pressure absorption, climatising, comfortable
  • Benefit for joints, musculature, ligaments and spine
Specifications
Specifications
Gray
Textile
3574