Learn More
Binder Installation CO.2 Control Module
Product Code. 30503386 Shop All BINDER Products
Click to view available options
Duration:
1 Day
Unit Size:
1 unit
Product Code. 30503386
Brand: Binder DL100130
Installation of the BINDER CO2 control module (0 to 1% or 0 to 20%)
  • Mount the supply box, insert the CO2 sensor and the gas hose into the interior, seal the access port, connect the box to an existing CO2 gas supply, update the controller data set, and check function
Specifications
For Use With (Equipment) BINDER KB PRO/KBF/KBF PRO and CO2 Gassing Module
Duration 1 Day
Product Type Installation Remote Support