Learn More
Binder Installation CO.2 Control Module
Shop All BINDER Products
Click to view available options
Duration:
1 Day
Unit Size:
1 unit
Description
- Mount the supply box, insert the CO2 sensor and the gas hose into the interior, seal the access port, connect the box to an existing CO2 gas supply, update the controller data set, and check function
Specifications
Specifications
| For Use With (Equipment) | BINDER KB PRO/KBF/KBF PRO and CO2 Gassing Module |
| Duration | 1 Day |
| Product Type | Installation Remote Support |