Learn More
Novus Biologicals™ InstantBlue Protein Stain
Product Code. 18783464 Shop All Bio Techne Products
Click to view available options
Quantity:
1000 mL
Unit Size:
1 L
Product Code. 18783464
Brand: Novus Biologicals™ ISB1L
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
InstantBlue is a ready to use Coomassie protein stain for polyacrylamide gels.
  • Ultra-fast staining – Results in 15 minutes or less
  • Single step procedure – No washing, fixing, microwaving or destaining
  • High sensitivity – 5ng bands detectable
  • Clear background – High signal/noise ratio
  • Flexible – No over staining
  • Quantitative – Batch to batch consistency
  • Efficient – Only 25 mL per gel
  • Safe composition – Non-toxic. No fume hood or solvent disposal required
  • Methanol free – No gel shrinkage or protein methylation
  • Acetic acid free – No protein aceylation
  • MS Compatible 1 – Destainable. No residual methylation or acetylation
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 4°C
Quantity 1000 mL