missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JULABO™ Insulation for Reinforced Tubing
Insulation for Reinforced Tubing is for use with 1.5 in. ID reinforced tubing.
Brand: JULABO™ 8930448
Additional Details : Weight : 0.20000kg
Description
- 48 mm inner diameter
Specifications
Insulation | |
Reinforced Tubing 1.5 in. ID | |
1 m | |
48 mm ID |