Learn More
Grant Instruments™ Interchangeable Block for QB Series Dry Block Heaters
Product Code. p-8014622
Click to view available options
Tube Size:
0.2mL
0.5mL
1.5mL
10mm
12mm
13mm
16.7 mm
16mm
17mm
18mm
2.0mL
24mm
50mL
None
No. of Places:
10
12
24
4
5
56
Solid Block
Depth (Metric):
14 mm hole depth
30 mm hole depth
35 mm hole depth
50 mm hole depth
53.5 mm hole depth
75 mm hole depth
None
Unit Size:
Each
Product Code. 11490908
Brand: Grant Instruments™ QB0
Only null left
Add to basket
Edge
Add to basket
For use with 0.2mL tubes, strips and 96-well microtitre plates used in molecular biology and biotechnology applications. Grant Instruments™ Interchangeable Block for QB Series Dry Block Heaters can withstand temperatures up to 100°C.
  • For use with QBD1, QBD2, QBD4, QBH2, QBA1 and QBA2 dry block heating system
  • Accommodates a variety of test tube sizes
  • Available in plain block format without holes
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
No. of Places Solid Block
Type Block
Tube Size None
Dimensions (L x W x H) 140 x 50 x 63 mm
Depth (Metric) None