Learn More
Fisherbrand™ Interchangeable Dry Bath Block
Product Code. 17501896 Shop All Fisherbrand Products
Click to view available options
No. of Places:
0
4
5
10
12
24
56
96
Tube Size:
0 mm
0.2 mL
0.5 mL
10 mm
12 mm
13 mm
14 mm
1.5 mL
16 mm
17 mm
18 mm
2 mL
24 mm
5 mL
50 mL
Unit Size:
Each
Product Code. 17501896
Only null left
Add to basket
Add to basket
For use with Fisherbrand™ Dry Block Heaters.
  • Aluminum construction provides fast heat-up
TRUSTED_SUSTAINABILITY
Specifications
No. of Places 24
Type Block
For Use With (Application) Falcon Tubes
Tube Size 0.5 mL