Learn More
Memmert™ Interior Lighting for UN30
Product Code. 17510156 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN30 Oven
Unit Size:
Each
Product Code. 17510156
Brand: Memmert™ UN30R0
Only null left
Add to basket
Add to basket
Interior lighting for observing the load.
  • Accessory
  • Interior Lighting
Specifications
For Use With (Equipment) UN30 Oven
Type Interior Lighting