Learn More
Kern Sohn Internal Rechargeable Battery Pack KB-A01N
Product Code. 17331673
Encompass_Preferred
Click to view available options
:
Balance Accessory
Regulatory: This product is under eco-contribution tax.
Product Code. 17331673
Brand: Kern Sohn KBA01N
Regulatory: This product is under eco-contribution tax.
Rechargeable battery pack for use with bench scales.
  • Rechargeable battery pack internal for counting scales KERN CDS and KERN CKE (models with weighing plate size W x D 150 x 170 mm) and precision balances KERN KB and KERN PKS and platform scale num/1] KERN DS
Specifications
For Use With (Equipment) Bench Scales
Type Rechargeable Battery Pack