missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Intra Acrosomal Protein Overexpression Lysate

Product Code. 18027969 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18027969 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18027969 Supplier Novus Biologicals™ Supplier No. NBP210987

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The Intra Acrosomal Protein Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Cell Line The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_163. The protein contains a C-terminal DDK tag.
For Use With (Application) Western Blot
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens Intra Acrosomal Protein, mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.