Learn More
Invitrogen™ ChargeSwitch™-Pro PCR Clean-Up Kit
GREENER_CHOICE
Product Code. p-4926420
Click to view available options
No. of Reactions:
10 Preps
250 Preps
50 Preps
Unit Size:
Each
Product Code. 10357943
Brand: Invitrogen™ CS32010
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Includes
Kit contains columns, collection tubes, recovery tubes, binding buffer, wash buffer, and elution buffer.
The ChargeSwitch™-Pro PCR Clean-up Kit purifies high-quality DNA fragments without the use of ethanol or harsh salts known to interfere with enzymatic reactions.
The ChargeSwitch™-Pro PCR Clean-up Kit purifies high-quality DNA fragments without the use of ethanol or harsh salts known to interfere with enzymatic reactions.
• 100% ethanol-free, guanidinium-free, organic solvent-free protocols
• Improved downstream performance
• Complete kit for fast, hassle-free setup
• 20% fewer handling steps than most PCR clean-up protocols
• Purification of up to 500 μL of PCR product in one column
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage
The ChargeSwitch™-Pro PCR Clean-up Kit contains columns, collection tubes, recovery tubes, binding buffer, wash buffer, and elution buffer.
Store at room temperature. Guaranteed stable for 6 months when properly stored.
Format Kit
Green Features Fewer resources used and less waste, Less hazardous
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line ChargeSwitch
Product Type PCR Clean-Up Kit
Quantity 10 Preps
Shipping Condition Room Temperature
Target DNA from Enzymatic Reactions (e.g. PCR)
For Use With (Application) Next-Generation Sequencing (NGS), Cloning, Standard PCR
Isolation Technology ChargeSwitch™ Chemistry
No. of Reactions 10 Preps
Sample Type PCR Products