missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™ Purification Buffer (N5)

Product Code. 10397893
Click to view available options
Quantity:
300 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10397893

Brand: Invitrogen™ CS11210

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Scalable for single-tube use or for automated extraction

  • ChargeSwitch Forensic DNA Purification Kit is magnetic bead method
  • Designed to accommodate all forensic crime scene sample types for STR profiling
  • Scalable for single-tube use or for automated extraction

DNA & RNA Purification & Analysis, DNA Extraction, Genomic DNA Purification, gDNA from Forensic Samples

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Buffer Binding Buffer
Quantity 300 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.