missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Go! Agarose Gels, 2%

Product Code. p-4926741
Click to view available options
Quantity:
10 Gels/Pk.
20 Gels/Pk.
Unit Size:
Each
Pack of 20
This item is not returnable. View return policy

Product Code. 12393643

Brand: Invitrogen™ G441002

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

For performing agarose gel electrophoresis with only a few samples to analyze

  • Precast agarose gel cassettes each have four wells and are premixed with a blue light excitable nucleic acid stain
  • Can be stored at room temperature and are ready to use without additional buffers or reagents
  • Designed specifically to work with the components of the E-Gel Go! System
  • For a quick check of end point PCRs, restriction digests, plasmid preps or other molecular biology reactions
  • Use proprietary stain formulated to provide the high detection sensitivity and can be visualized with traditional UV transilluminators
  • When using blue light the bands must be viewed through an amber filter
  • Sharp bands and repeatable, reliable results
  • Single comb well format
  • Sample Volume; 10μL

  • For linear double stranded DNA fragments ranging from 1.5kbp to 100bp in size the 2% gels offer excellent resolution and separation

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Compatible Ladders E-Gel™ 1 Kb Plus Express Ladder
Content And Storage • Each pack contains 10 individually sealed 2% E-Gel™ Go! precast agarose gel cassettes

Store at room temperature.
Gel Percentage 2%
Gel Type E-Gel
Product Type Go Agarose Gel
Sample Loading Volume 10 μL
Separation Range 50 to 2000 bp
Stain Type SYBR™ Gold
Wells 4-well
Quantity 10 Gels/Pk.
Shipping Condition Room Temperature
System Type E-Gel™ Go!
Well Design Single Comb
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.