Learn More
Invitrogen™ Gateway™ pYES-DEST52 Vector
Designed for rapid cloning with Gateway entry clone using lambda phage site-specific recombination and subsequent expression in Saccharomyces cerevisiae
Brand: Invitrogen™ 12286019
669.06 EUR valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Additional Details : Weight : 0.02000kg
Includes: The pYES2-DEST52 Gateway destination vector is provided supercoiled and lyophilized.
Description
- As part of YESVector Collection, pYES2-DEST52 carries promoter and enhancer sequences from GAL1 gene for regulated expression in S.cerevisia
- 2μ origin of replication for high-copy maintenance
- Gene for auxotrophic selection on economical minimal medium
- C-terminal V5 epitope and polyhistidine(6xHis) tags for efficient detection and purification
- tR sites for efficient recombination with any attL-flanked Gateway entry vector
Cloning, Gateway Cloning, Protein Expression, Proteins, Expression, Isolation and Analysis, S. cerevisiae Expression Systems, Yeast Expression
Specifications
GAL1 | |
The pYES2-DEST52 Gateway™ destination vector is provided supercoiled and lyophilized. Store at -20°C. The vector is guaranteed stable for 6 months when properly stored. | |
His Tag (6x), V5 Epitope Tag | |
pYES, pDEST |
Gateway System Destination Expression Vector | |
Ampicillin (AmpR) | |
Gateway | |
Gateway™ |
For Research Use Only. Not for use in diagnostic procedures.