missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ 384 Pre-Lysis Buffer

Up to 96 plasmid DNA samples can be simultaneously purified

Brand:  Invitrogen™ 12263034

Additional Details : Weight : 0.45000kg

Product Code. 11508696

  • 291.00€ / 125mL
Estimated Shipment: 07-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: 125mL bottle

Description

Description

The PureLink 384 Plasmid Purification Plate is designed for ultra high-throughput DNA isolation. By utilizing same technology as PureLink 96 Plasmid Purification System, it reduces time and effort while reliably isolating high quality plasmid DNA for DNA sequencing, PCR, and restriction digests.

  • PureLink 96: Sample size 1.3mL culture; Typically yields up to 3-5μg
  • PureLink 384: Sample size 200μL culture; Typically yields up to 2-3μg
  • Purify 96 or 384 plasmid DNA samples simultaneously
  • Unique centrifugation method eliminates cross-contamination
  • PureLink 96 Plasmid Purification System is designed for high-throughput isolation of high copy number plasmids
  • Novel technology eliminates time-consuming cell harvesting and alkaline Lysis steps, saving time and effort
  • Unique centrifugation-based format of the PureLink 96 Plasmid Purification System eliminates problematic cross-contamination between wells, a critical problem in PCR and microarray analysis
  • Isolated plasmid DNA is ideal for use in automated sequencing reactions, offering superior read lengths and data quality
  • Samples may be processed 96 at time or in smaller increments
  • PureLink 96 Plasmid Purification System is compatible with variety of centrifuges, robots, pipetting devices, automated sequencers, and sequencing chemistries

Automated Nucleic Acid Purification, Automated Plasmid DNA Purification, DNA & RNA Purification & Analysis, DNA Extraction, Miniprep, Plasmid DNA Purification

Specifications

Specifications

Lysis Buffer
125 ml The PureLink™ 96 Components are shipped at room temperature. Upon receipt, store the components at room temperature.
Automated Sequencing Reaction
PureLink™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.