Learn More
Invitrogen™ PureLink™ Viral Lysis Buffer
Product Code. 10725403
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 10725403
Brand: Invitrogen™ 12282500
Includes
Filter Plate, Receiver plates, deep-well blocks, foil tape, carrier RNA, proteinase K, Viral Lysis Buffer, Wash Buffer, and RNase-free water.
PureLink 96 Viral RNA/DNA Kit is 96-well silica plate system that can extract both viral RNA and DNA from 200-μL cell-free fluids which uses low-depth 96-well centrifuge bucket for ease-of-use.
  • PureLink 96 Viral RNA/DNA Kit is 96-well silica plate system that can extract both viral RNA and DNA from 200-μL cell-free fluids which uses low-depth 96-well centrifuge bucket for ease-of-use
  • Provide flexibility in centrifuge choice. Minimum speeds for 96 well processing of 2250 x g and rotor bucket depth of 5 cm make this kit one of the most adaptable systems for existing laboratory centrifuges
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Buffer Lysis Buffer
Content And Storage The PureLink™ 96 Viral RNA/DNA Kit contains a PureLink™ Filter Plate, Receiver plates, deep-well blocks, foil tape, carrier RNA, proteinase K, Viral Lysis Buffer, Wash Buffer, and RNase-free Water. The kit is shipped and stored at room temperature and the lyophilized tRNA should be removed and stored at -20°C upon receipt. The proteinase K should be stored at +4°C.
For Use With (Application) Nucleic Acid Extraction
Product Line PureLink
Product Type Viral Lysis Buffer
Quantity 500 mL
Shipping Condition Room Temperature