Learn More
Invitrogen™ ThermoScript™ RT-PCR System for First-Strand cDNA Synthesis
Click to view available options
:
25 reactions
Specifications
Specifications
| Content And Storage | 25μL ThermoScript RT (15U/μL); 500μL 5X cDNA Synthesis Buffer; 250μL 0.1M DTT; 100μL 10mM dNTP Mix; 25μL RNaseOUT (40U/μL); 25μL Oligo (dT)20 (50μM); 50μL Random Hexamers (50ng/μL); 1.25mL DEPC-Treated Water; 50μL E. coli RNase H (2U/μL). Store at -20deg.C |
| Primer | Random Primers, Oligo dT Primers |
| Quantity | 25 reactions |
| For Use With (Application) | cDNA Synthesis, RT-PCR |