missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Guanidine Isothiocyanate

UltraPure Guanidine Isothiocyanate is a strong protein denaturant when used at high concentrations.

Brand:  Invitrogen™ 15535016

524.02 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.



Additional Details : Weight : 0.39000kg

Product Code. 11578746

  • 786.00€ / 500g
Estimated Shipment: 15-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

UltraPure Guanidine Isothiocyanate is a strong protein denaturant when used at high concentrations. It is useful in the isolation of intact RNA from sources rich in RNase such as pancreas.

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Store at room temperature.
Guanidine Isothiocyanate
DNase-Free, RNase-Free
Room Temperature
RNA Extraction
UltraPure™
500 g
Powder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.