Learn More
Invitrogen™ Zero Background™ /Kan Cloning Kit
Product Code. 11544946
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
Product Code. 11544946
Brand: Invitrogen™ K260001
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Includes
25μg supercoiled pZErO-1 (the Zero Background Cloning Kit)or pZErO-2 vector (the Zero Background/Kan Cloning Kit) in TE, 200ng blunt-end test inserts, TOP10 E. coli stab, 25μL T4 DNA ligase, 10X ligation buffer, 1mL sterile water, and 2mL sterile TE. The Zero Background Cloning Kit also contains 1.25mL Zeocin selection agent.
Includes pZErO vector with Zeocin or kanamycin resistance for selection in E. coli
  • pZErO vectors use positive selection to eliminate high background
  • pZErO-1 vector takes advantage of small size of Zeocin resistance gene to reduce the overall vector size for increased ligation and transformation efficiencies
  • pZErO-2 vector includes the kanamycin resistance gene
Cloning, PCR Cloning
TRUSTED_SUSTAINABILITY
Specifications
Bacterial or Yeast Strain TOP10
Content And Storage Store TOP10 stab at room temperature. Store the remaining kit components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
Quantity 25 μg
Vector TA Cloning Vectors, pCR
For Research Use Only. Not for use in diagnostic procedures.