missing translation for 'onlineSavingsMsg'
Learn More

Iodine Indicator, Technical, Indicator Grade, Fisher Chemical™

Catalog Number 12627915
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
3703398
This item is not returnable. View return policy
3703398

Iodine Indicator, Technical, Indicator Grade

TRUSTED_SUSTAINABILITY
Packaging Plastic powder jar
Color White
pH 5 to 7
Quantity 1 kg
Physical Form Powder
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'