missing translation for 'onlineSavingsMsg'
Learn More

Iodine solution, 0.5M EU Pharmacopoeia, Fisher Chemical™

Catalog Number 12930864
Click to view available options
Quantity:
1000 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Concentration 0.5M
CAS 7732-18-5
Packaging Glass Bottle
Quantity 1000 mL
Grade Eu. Ph. Reference
Chemical Name or Material Iodine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'