missing translation for 'onlineSavingsMsg'
Learn More

Fluoride Ionic Strength Adjuster, TISAB3, Reagecon™

Product Code. 11718495 Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 11718495

Brand: Reagecon TISAF5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for fluoride determination in the presence of Al(III) using an Ion Selective Electrode (ISE). Fluoride Ionic Strength Adjuster, TISAB3, Reagecon™ is used to measure ion activity when the sample solution needs additional removal of interfering ions and/or pH buffering.

An Ionic Strength Adjuster (ISA) is used to level the ion activity for all sample solutions to enable ionic activity measurements in sample solutions using an Ion Selective Electrode (ISE). If the sample solution needs additional removal of interfering ions and/or pH buffering, it is not called ISA solution, but a TISAB (total ionic strength adjustment buffer) solution.

  • The TISAB keeps the ionic strength of the sample and standard high, constant and similar, which enables activity measurement to be read as concentration
  • The TISAB keeps the pH value constant, which helps suppress or eliminate interfering ions
  • The TISAB eliminates any matrix, hysteresis or erroneous liquid junction potentials that might affect the accuracy of the test result
  • A TISAB should never react with the sample or standard chemically

Specifications

Boiling Point 100°C
For Use With (Equipment) Ion Selective Electrode
pH 5 to 5.5
Physical Form Liquid
Quantity 500 mL
Formulation Potassium Hydrogen Phthalate, Potassium Dihydrogen Phosphate and Disodium Hydrogen Phosphate
Traceability to NIST Yes
Vapor Pressure 16hPa at 20°C
compliance-icons compliance-icons
Product Identifier
  • FLUORIDE TISAB 3 TOTAL ISA SOLUTION (CONCENTRATE)Addition rate 5ml per 50ml sample
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : acetic acid, ammonium chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.