missing translation for 'onlineSavingsMsg'
Learn More

Melford IPTG

Product Code. p-200087077
Click to view available options
Quantity:
1 g
5 g
25 g
50 g
100 g
Unit Size:
100g
1g
25g
50g
5g
This item is not returnable. View return policy

Product Code. 30018359

Brand: Melford I5600025.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

IPTG is combined with X-GAL to detect lac+ colonies or cells in a colorimetric assay.

IPTG is combined with X-GAL to detect lac+ colonies or cells in a colorimetric assay. Distinguishes recombinants (white) from non- recombinants (blue) in cloning strategies using vectors such as Lambda-11, M13mp 18 and 19, pUC18 and 19, pUR222 containing the lacZ gene.

Dioxane-Free

Water Content: <0.49%

Specifications

CAS 367-93-1
Molecular Formula C9H18O5S
Quantity 25 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.