Learn More
Iron for AAS 1g/L HNO3 matrix, Fisher Chemical™
Catalog Number 12923275
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Concentration 1g/L
Quantity 500 mL
Solution Contains HNO3