missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(II) sulfate heptahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.5-104.5% (manganometric), Honeywell Fluka™
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 250 g |
Linear Formula | FeSO4 · 7H2O |
Formula Weight | 278.01g/mol |
Percent Purity | 99.5 to 104.5% (manganometric) |
Grade | puriss. |
Packaging | Plastic Bottle |
Vapor Pressure | 14.6mmHg (25°C) |
Density | 1.898g/mL at 25°C |
Chemical Name or Material | Iron(Ii) Sulfate Heptahydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'