missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) sulfate heptahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.5-104.5% (manganometric), Honeywell Fluka™
BIOPHARMA

Catalog Number 15651140
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
7782-63-0
FeH14O11S
278.01
MFCD00149719
SURQXAFEQWPFPV-UHFFFAOYSA-L
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
62662
CHEBI:75836
iron(2+);sulfate;heptahydrate
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
7782-63-0
FeH14O11S
278.01
MFCD00149719
SURQXAFEQWPFPV-UHFFFAOYSA-L
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
62662
CHEBI:75836
iron(2+);sulfate;heptahydrate
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, 99.5-104.5% (manganometric)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 4 x 5 kg
Linear Formula FeSO4 · 7H2O
Formula Weight 278.01
Percent Purity 99.5 to 104.5% (manganometric)
Grade puriss.
Vapor Pressure 14.6mmHg (25°C)
Density 1.898g/mL at 25°C
Chemical Name or Material Iron(Ii) Sulfate Heptahydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'