missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) 1,1,1-trifluoro 2,4-pentanedionate, Thermo Scientific Chemicals

Catalog Number p-7050675
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14526-22-8
C15H12F9FeO6
515.09
MFCD00017760
PDLKINIVWBCDRU-UHFFFAOYSA-N
ferric iron 1,1,1-trifluoro-4-oxopent-2-en-2-olate
71299627
[Fe+3].CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14526-22-8
C15H12F9FeO6
515.09
MFCD00017760
PDLKINIVWBCDRU-UHFFFAOYSA-N
ferric iron 1,1,1-trifluoro-4-oxopent-2-en-2-olate
71299627
[Fe+3].CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F

It is used as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 110°C to 112°C
Quantity 5 g
Solubility Information Insoluble in water.
IUPAC Name iron(3+);(Z)-1,1,1-trifluoro-4-oxopent-2-en-2-olate
Formula Weight 515.09
Physical Form Crystalline
Chemical Name or Material Iron(III) 1,1,1-trifluoro 2,4-pentanedionate
compliance-icons
Product Identifier
  • Iron(III) 1,1,1-trifluoro 2,4-pentanedionate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'