missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka™ Iron(Iii) Nitrate Nonahydrate, Honeywell Fluka™

ACS Reagent,≥98%

Brand:  Honeywell Fluka™ 216828-1KG

Additional Details : CAS : 7782-61-8 Poids : 1.00000kg Transport : UN number : 3260 Chem class : 8 Pack group : II

Quantity 1kg

 View more versions of this product

Product Code. 15662040

  • 131.10€ / 1kg


Expédition estimée
12-10-2020
Add to basket

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7782-61-8
403.992
SZQUEWJRBJDHSM-UHFFFAOYSA-N
16211566
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Fe+3]
FeH18N3O18
MFCD00149708
ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
iron(3+);trinitrate;nonahydrate

Specifications

Iron(Iii) Nitrate Nonahydrate
FeH18N3O18
MFCD00149708
ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Fe+3]
403.992
404g/mol
ACS Reagent
7782-61-8
Fe(NO3)3 · 9H2O
3260
SZQUEWJRBJDHSM-UHFFFAOYSA-N
iron(3+);trinitrate;nonahydrate
16211566
≥98%
1kg
Videos
Documents