missing translation for 'onlineSavingsMsg'
Learn More

Iron(Iii) Nitrate Nonahydrate, Honeywell Fluka™

Catalog Number p-7071884
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
2.5 kg
This item is not returnable. View return policy
7782-61-8
FeH18N3O18
403.99
MFCD00149708
SZQUEWJRBJDHSM-UHFFFAOYSA-N
ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
16211566
iron(3+);trinitrate;nonahydrate
This item is not returnable. View return policy
7782-61-8
FeH18N3O18
403.99
MFCD00149708
SZQUEWJRBJDHSM-UHFFFAOYSA-N
ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
16211566
iron(3+);trinitrate;nonahydrate

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 100 g
Linear Formula Fe(NO3)3 · 9H2O
UN Number 3260
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Formula Weight 404g/mol
Percent Purity ≥98%
Grade ACS Reagent
Chemical Name or Material Iron(Iii) Nitrate Nonahydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'