missing translation for 'onlineSavingsMsg'
Learn More

Iron, Technical, Filings, Fine (Metal), Fisher Chemical™

Product Code. 10090180 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 g
1 kg
3 kg
Unit Size:
1kg
250g
3kg
This item is not returnable. View return policy

Product Code. 10090180

Brand: Fisher Chemical I/0850/60

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Color Gray
Melting Point 1535°C
Boiling Point 3000°C
Quantity 1 kg
Packaging Plastic powder jar
Particle Size 0.3 to 0.72mm
Vapor Pressure 1mmHg 1787
Form Solid
Formula Weight 55.84g/mol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.