missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ISO 7010 Sign - Wear ear protection

Prevent costly accidents with ISO 7010 safety signs for optimal impact and recognizability.

Brand:  Brady™ 818277

Additional Details : Poids : 0.00830kg

 View more versions of this product

Product Code. 15664447

  • 26.10€ / Each


Expédition estimée
30-09-2020
Add to basket

Description

Description

  • Polypropylene
  • For indoor and outdoor use
Specifications

Specifications

ISO 7010
Global
1
315 mm
1.501 mm
White on Blue
Polypropylene
Circle
Indoor and Outdoor
-40°C to 120°C
Videos
Documents