Learn More
Brady™ ISO Safety Sign - Disinfect Your Hands, English
Product Code. 30280561
Click to view available options
Dimensions (L x W):
150 x 50 mm
148 x 210 mm
210 x 297 mm
297 x 105 mm
297 x 420 mm
450 x 150 mm
600 x 200 mm
Material:
Aluminum
Laminated Polyester
Polypropylene
Unit Size:
Each
Product Code. 30280561
Brand: Brady™ 322258
Only null left
Add to basket
Add to basket
Sign to signify that hands must be disinfected. Text is in English.
  • Sign compliant to ISO 7010
Spécification
Certifications/Compliance ISO 7010; Compliant with Directive 2011/65/EU and 2015/863
Alternate Part Number M/M061/EN593/ALU-297x420-1
Grade M061
Material Aluminum
Quantity 1 Ea.
Mounting Type Surface Mount, Wall Mount
For Use With (Application) Facility Identification, Safety Identification
Height (English) 16.54 in.
Height (Metric) 420 mm
Thickness 1 mm
Width (English) 11.69 in.
Width (Metric) 297 mm
Temperature Range (Metric) -40°C to +140°C
Shape Rectangular
Temperature Range (English) -40°F to +284°F
Languages English
Dimensions (L x W) 297 x 420 mm
Displayed Text Disinfect your hands
Product Type Personal Hygiene Sign
Color White on Blue