Learn More
Brady™ ISO Safety Sign, Floor Marking - Disinfect Your Hands
Product Code. p-200109841
Click to view available options
Languages:
Danish
Dutch
English
Finnish
French
German
Italian
Norwegian
Portuguese
Spanish
Swedish
Turkish
Unit Size:
Each
Product Code. 30280498
Brand: Brady™ 323394
Only null left
Add to basket
Add to basket
Floor sign to signify that hands must be disinfected.
  • Sign compliant to ISO 7010
Specifications
Certifications/Compliance ISO 7010; Compliant with Directive 2011/65/EU and 2015/863
Alternate Part Number M/M061/EN593/FLO-DIA350-1
Grade M061
Material Polyester
Quantity 1 Ea.
Diameter (Metric) 350 mm
Mounting Type Surface Mount, Wall Mount
For Use With (Application) Floor Marking
Thickness 0.28 mm
Shape Round
Languages English
Displayed Text STOP Disinfect your hands
Product Type Personal Hygiene Sign
Color White on Blue