Learn More
Alla France™ ISO Standard Hydrometer
Click to view available options
Density (Metric):
0.130 to 0.140g/mL
0.900 to 0.100g/mL
1000 to 1050g/L
1000 to 1100g/L
1100 to 1150g/L
1100 to 1200g/L
1200 to 1300g/L
950 to 1000g/L
Length (Metric):
250 mm
270 mm
Unit Size:
Each
Description
- Density Hydrometers (g/mL)
- ISO, BS, DIN, NF... MRA Traceable
- General Purpose at 20°C
- The scale of the hydrometer is fully marked with all the reference norms, the calibration temperature, the surface tension, the individual number of the instrument, the serial number and the ALLA FRANCE logo
- Reading below meniscus.
- Systematically delivered with a Traceability and Conformity Certificate.
- On request, can be supplied with "EU" Traceable Calibration Certificate or "EO" Official Calibration Certificate
- from 0.600 to 1.000 g/mL= 16 at 35 mN/m (low tension)
- from 1.000 to 1.300 g/mL= 55 mN/m (medium tension)
- from 1.300 to 2.000 g/mL= 75 mN/m (high tension)
Specifications
Specifications
| Quantity | 1Each |
| Type | ISO Standard Hydrometer |
| Length (Metric) | 250 mm |
| Density (Metric) | 1000 to 1100g/L |
| Specific Gravity Range | 1000 to 1100 g/L |