Learn More
Alla France™ ISO Standard Hydrometer
Product Code. 11743423
Click to view available options
Density (Metric):
0.130 to 0.140g/mL
0.900 to 0.100g/mL
1000 to 1050g/L
1000 to 1100g/L
1100 to 1150g/L
1100 to 1200g/L
1200 to 1300g/L
950 to 1000g/L
Length (Metric):
250 mm
270 mm
Unit Size:
Each
Product Code. 11743423
Brand: Alla France™ 3100MC120/20/Mqp
Only null left
Add to basket
Add to basket
Use these hydrometers for a variety of applications. Alla France™ ISO Standard Hydrometer feature a benchmark engraved on the stem to show any displacement of the scale.
  • Density Hydrometers (g/mL)
  • ISO, BS, DIN, NF... MRA Traceable
  • General Purpose at 20°C
  • The scale of the hydrometer is fully marked with all the reference norms, the calibration temperature, the surface tension, the individual number of the instrument, the serial number and the ALLA FRANCE logo
  • Reading below meniscus.
  • Systematically delivered with a Traceability and Conformity Certificate.
  • On request, can be supplied with "EU" Traceable Calibration Certificate or "EO" Official Calibration Certificate
Calibrated according to the following surface tensions:
  • from 0.600 to 1.000 g/mL= 16 at 35 mN/m (low tension)
  • from 1.000 to 1.300 g/mL= 55 mN/m (medium tension)
  • from 1.300 to 2.000 g/mL= 75 mN/m (high tension)
Specifications
Quantity 1Each
Type ISO Standard Hydrometer
Length (Metric) 250 mm
Density (Metric) 1200 to 1300g/L
Specific Gravity Range 1200 to 1300 g/L