missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoguanine, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Isoguanine |
| CAS | 3373-53-3 |
| Molecular Formula | C5 H5 N5 O |
| Quantity | 250 mg |
| Synonym | 2H-Purin-2-one, 6-amino-3,7-dihydro-, 2H-Purin-2-one, 6-amino-1,3-dihydro- (9CI), Isoguanine (6CI,7CI,8CI), 6-Amino-3,7-dihydro-2H-purin-2-one, 2-Hydroxy-6-aminopurine, 2-Hydroxyadenine, 2-Oxoadenine, 2-Oxyadenine, 6-Amino-2-hydroxypurine, Guanopterin, NSC 241501, 6-Amino-7H-purin-2-ol) |
| InChI Formula | InChI=1S/C5H5N5O/c6-3-2-4(8-1-7-2)10-5(11)9-3/h1H,(H4,6,7,8,9,10,11) |
| SMILES | NC1=NC(=O)Nc2nc[nH]c12 |
| IUPAC Name | 6-amino-3,7-dihydropurin-2-one |
| Molecular Weight (g/mol) | 151.13 |
| Formula Weight | 151.0494 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction