missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-8-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4228755
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
61563-43-7
C10H7NO2
173.17
MFCD06738935
VMNZQPRIUSJCOH-UHFFFAOYSA-N
8-isoquinolinecarboxylic acid, 8-carboxyisoquinoline, 8-isoquinolinecarboxylicacid, 8-carboxy-2-azanaphthalene
21393830
isoquinoline-8-carboxylic acid
C1=CC2=C(C=NC=C2)C(=C1)C(=O)O
This item is not returnable. View return policy
61563-43-7
C10H7NO2
173.17
MFCD06738935
VMNZQPRIUSJCOH-UHFFFAOYSA-N
8-isoquinolinecarboxylic acid, 8-carboxyisoquinoline, 8-isoquinolinecarboxylicacid, 8-carboxy-2-azanaphthalene
21393830
isoquinoline-8-carboxylic acid
C1=CC2=C(C=NC=C2)C(=C1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 263.0°C to 267.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 173.17
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Isoquinoline-8-carboxylic acid
compliance-icons
Product Identifier
  • Isoquinoline-8-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'