missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline N-oxide, 98%, Thermo Scientific™

Catalog Number 11408627
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
1532-72-5
C9H7NO
145.161
MFCD00006903
RZIAABRFQASVSW-UHFFFAOYSA-N
isoquinoline n-oxide, isoquinoline 2-oxide, isoquinoline-n-oxide, isoquinoline oxide, isoquinoline, 2-oxide, isoquinolin-n-oxide, isoquinolin-2-ium-2-olate, isoquinolinen-oxide, acmc-1boxv, 2-oxido-isoquinolin-2-ium
290378
2-oxidoisoquinolin-2-ium
C1=CC=C2C=[N+](C=CC2=C1)[O-]
This item is not returnable. View return policy
1532-72-5
C9H7NO
145.161
MFCD00006903
RZIAABRFQASVSW-UHFFFAOYSA-N
isoquinoline n-oxide, isoquinoline 2-oxide, isoquinoline-n-oxide, isoquinoline oxide, isoquinoline, 2-oxide, isoquinolin-n-oxide, isoquinolin-2-ium-2-olate, isoquinolinen-oxide, acmc-1boxv, 2-oxido-isoquinolin-2-ium
290378
2-oxidoisoquinolin-2-ium
C1=CC=C2C=[N+](C=CC2=C1)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 103°C to 105°C
Boiling Point 170°C (3 mmHg)
Quantity 5 g
Beilstein 114345
Sensitivity Hygroscopic
Formula Weight 145.16
Percent Purity 98%
Chemical Name or Material Isoquinoline N-oxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'