Learn More
Fisherbrand™ Isotemp™ Heated Bath Circulators: Model 5150 H24
Product Code. p-8382559 Shop All Fisherbrand Products
Click to view available options
Frequency:
50 Hz
Unit Size:
Each
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11376671
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Reliable, high-wattage heaters that deliver rapid fluid heat up and ensure fluid compatibility for your application.
Standard high temperature cutout (HTC) circuitry is user-selectable to ensure application temperatures do not exceed set point parameters
  • High temperature and low level cut-outs
  • Compact footprint maximizes bench space
  • Includes external circulation plumbing with 8-mm and 12-mm fittings and supply and return clamps
  • Work area cover included on all heated baths
  • High quality stainless steel baths
  • All bath drains are located on the front of the unit with an integrated valve
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 15.4 to 19.6 L
Pump Type Force
Temperature Range 10°C to 150°C
Heating Capacity 1200 w
Dimensions (L x W x H) 56.13 x 36.32 x 39.62 cm
Length (Metric) 56.13 cm
Width (Metric) 36.32 cm
Width (Metric) Interior 29.46 cm
Height (Metric) 39.62 cm
Weight (Metric) 15.9 kg
Electrical Requirements 230 V 50 Hz
Voltage 230 V
Frequency 50 Hz