Learn More
Eppendorf™ IsoTherm™ System Starter Set
Product Code. 10114721 Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
0.5mL tubes
1,5/2,0mL tubes
Unit Size:
Each
Product Code. 10114721
Brand: Eppendorf™ 3880000011
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Holds up to 24 micro tubes. The IsoTherm-System is the ideal system for safely cooling your samples in 0.5mL or 1.5 mL/2.0 mL tube formats at -21°C or 0°C throughout your entire lab workflow.
  • IsoPack™ (cold pack) white for 0°C and blue for -21°C, cooled to refrigeration temperature in freezer compartment
  • Isothermal times of >6hr (0°C, white) and >3hr (-21°C, blue)
  • No more melting ice baths (danger of contamination)
  • All system components can be used separately
  • Racks are stackable, autoclavable, markable, floatable and can be centrifuged in a microtitre-plate rotor
  • Specializes in cooling samples effectively and constantly at -21°C or 0°C over many hours.
TRUSTED_SUSTAINABILITY
Specifications
Type IsoTherm-System Starter Set
For Use With (Equipment) 0.5mL tubes