Learn More
Tocris Bioscience™ Ivosidenib
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Ivosidenib is a potent inhibitor of mutant isocitrate dehydrogenase enzyme 1 (mIDH1), with activity against a variety of mutant IDH1 enzymes (IC50 range = 5-19 nM). Reduces intracellular 2-HG (Cat. No. 6122) by 96% at 0.5 μM. Increases sensitivity of melanoma to Temozolomide (Cat. No. 2706) in vivo and inhibits wild-type IDH1 in low magnesium and low nutrient levels. Ivosidenib also inhibits growth of tumors in cell and animal models of pancreatic cancer and inhibits invasion and migration of chondrosarcoma cells in vitro.
Specifications
Specifications
| Chemical Name or Material | (2S)-N-{(1S)-1-(2-Chlorophenyl)-2-[(3,3-difluorocyclobutyl)amino]-2-oxoethyl}-1(4-cyanopyridin-2-yl)-N-(5-fluoropyridin-3-yl)-5-oxopyrrolidine-2-carboxamide |
| CAS | 1448347-49-6 |
| Quantity | 50 mg |
| Target | Isocitrate Dehydrogenase 1/IDH1 |
| Molecular Formula | C28H22ClF3N6O3 |