Learn More
GPE Scientific™ J Young Sample Flask with O-Ring Tap and Cold Finger Plain Arm
Product Code. 15874744
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Product Code. 15874744
Brand: GPE Scientific™ CSY/50
Only null left
Add to basket
Add to basket
GPE Scientific™ J Young Sample Flask with O-Ring Tap and Cold Finger Plain Arm is designed for use with Schlenk lines and a vacuum manifold and available in various capacities.
  • Designed with a side arm for evacuating or filling with gases
  • For connection to Schlenk lines
  • Includes O-ring tap
  • Includes cold finger plain arm
Specifications
Capacity (Metric) 50 mL
Includes O-ring tap and cold finger plain arm
Product Type Sample Flask